missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPV17 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10310-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
10310-1-AP is a rabbit polyclonal antibody raised against the full-length of human MPV17.
This gene encodes a mitochondrial inner membrane protein that is implicated in the metabolism of reactive oxygen species. Mutations in this gene have been associated with the hepatocerebral form of mitochondrial DNA depletion syndrome.Specifications
MPV17 | |
Polyclonal | |
Unconjugated | |
MPV17 | |
MPV17, Protein Mpv17, SYM1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17527, 4358 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P19258, P39210 | |
Mpv17 | |
MPV17 Fusion Protein Ag0312 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |