Läs mer
MR 121, Technical Grade, TRC
Produktkod. 30469847
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30469847 1 mg
30478433 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30469847 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM7501001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 185213-55-2
Molekylformel C24H28N3O3 . xHCl
Molekylvikt (g/mol) 406.5
Synonym 4-(11-Ethyl-3,4,8,9,10,11-hexahydrodipyrido[3,2-b:2',3'-i]phenoxazin-13-ium-1(2H)-yl)butanoate, MR 121, Technical Grade
LEDER OC(=O)CCCN1CCCc2cc3Nc4cc5CCCNc5cc4Oc3cc12
Specifikationer
Kemiskt namn eller material 1-(3-Carboxypropyl)-11-ethyl-3,4,8,9,10,11-hexahydro-2H-Dipyrido[3,2-b:2',3'-i]phenoxazin-13-ium inner salt
Kvantitet 1 mg
InChI formel InChI=1S/C24H27N3O3/c1-2-26-9-3-6-16-12-18-22(14-20(16)26)30-23-15-21-17(13-19(23)25-18)7-4-10-27(21)11-5-8-24(28)29/h12-15H,2-11H2,1H3/p+1