Läs mer
MRL-871, TRC
Produktkod. 30469413
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30469413 1 mg -
30473988 2.5 mg 2.5mg
30475781 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30469413 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM7502301MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1392809-08-3
Molekylformel C22H12ClF3N2O3
Molekylvikt (g/mol) 444.79
Synonym 4-[1-[2-Chloro-6-(trifluoromethyl)benzoyl]-1H-indazol-3-yl]benzoic Acid
IUPAC-namn 4-(1-(2-chloro-6-(trifluoromethyl)benzoyl)-1H-indazol-3-yl)benzoic acid
LEDER O=C(C1=CC=C(C2=NN(C3=C2C=CC=C3)C(C4=C(C(F)(F)F)C=CC=C4Cl)=O)C=C1)O
Specifikationer
Kemiskt namn eller material 4-[1-[2-Chloro-6-(trifluoromethyl)benzoyl]-1H-indazol-3-yl]benzoic Acid
Kvantitet 1 mg
InChI formel InChI=1S/C22H12ClF3N2O3/c23-16-6-3-5-15(22(24,25)26)18(16)20(29)28-17-7-2-1-4-14(17)19(27-28)12-8-10-13(11-9-12)21(30)31/h1-11H,(H,30,31)
Formel vikt 444.05