Learn More
MRP1 ABCC1 Mouse anti-Human, Unconjugated, Clone: 1G4A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67228-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The MRP1, ABCC1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MRP1, ABCC1. This antibody recognizes human antigen. The MRP1, ABCC1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
MRP1 ABCC1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot | |
1G4A2 | |
Unconjugated | |
150 μL | |
4363 | |
IgG1 |