Läs mer
MRP1 Antibody (QCRL), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB19291MFV610
Produktkod. 18515309
  • 6390.00 SEK / 0.10ml
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Beskrivning
Beskrivning
MRP1 Monoclonal antibody specifically detects MRP1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifikationer
Specifikationer
MRP1
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
0.1 mL
ABC Transporters, Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction
4363
Store at 4C in the dark.
Western Blot, Immunoprecipitation, Flow Cytometry, Immunocytochemistry, CyTOF
QCRL
Western Blot, Immunoprecipitation, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
ABC29, ABCC, ATP-binding cassette sub-family C member 1, ATP-binding cassette, sub-family C (CFTR/MRP), member 1, EC 3.6.3, EC 3.6.3.44, GS-X, Leukotriene C(4) transporter, LTC4 transporter, MRP1DKFZp781G125, MRPDKFZp686N04233, multidrug resistance associated protein 1, multidrug resistance protein, multidrug resistance-associated protein 1, multiple drug resistance protein 1, multiple drug resistance-associated protein
H69AR human small cell lung cancer cell line membrane extracts
RUO
Primary
Human
IgG1