missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
MRP4/ABCC4 Antibody (M4I-80), Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-42338
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MRP4/ABCC4 Monoclonal specifically detects MRP4/ABCC4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifikationer
| MRP4/ABCC4 | |
| Monoclonal | |
| Unconjugated | |
| ATP-binding cassette sub-family C member 4, ATP-binding cassette, sub-family C (CFTR/MRP), member 4, EST170205, MOAT-BbA464I2.1 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4), MOATBcanalicular multispecific organic anion transporter (ABC superfamily), MRP4MRP/cMOAT-related ABC transporter, multidrug resistance-associated protein 4, multispecific organic anion transporter B, Multi-specific organic anion transporter B | |
| Rat | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| The M4I-80 Mab was selected after immunization with a fusion protein containing the E. coli maltose binding protein and a fragment of the human MRP4 protein corresponding to amino acids 372-431. MRP4 transports cyclic nucleotides and anti-retroviral compounds. The M4I-80 Mab also reacts with the mouse orthologue of the transporter molecule (Mrp4). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| M4I-80 | |
| Western Blot 1:20-1:50, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:20-1:50, Immunohistochemistry-Frozen 1:20 | |
| ABCC4 | |
| The antibody reacts with the MRP4 antigen. | |
| 0.2 mL | |
| Lipid and Metabolism | |
| 10257 | |
| Human, Mouse | |
| Supernatant |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering