Learn More
MRP9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560239
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GPYLISDLDQ RGRRRSFAER YDPSLKTMIP VRPCARLAPN PVDDAGLLSF ATFSWLTPVM VKGYRQRLTV DTLPPLSTYD SSDTNAKRFR V Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.
MRP9 Polyclonal antibody specifically detects MRP9 in Human samples. It is validated for Immunohistochemistry.Specifications
MRP9 | |
Polyclonal | |
Unconjugated | |
ABCC12 | |
4930467B22Rik; Abcc12; ATP binding cassette subfamily C member 12; ATP-binding cassette protein C12 variant A; ATP-binding cassette sub-family C member 12; ATP-binding cassette transporter sub-family C member 12; ATP-binding cassette, subfamily C (CFTR/MRP), member 12; ATP-binding cassette, sub-family C (CFTR/MRP), member 12; MRP9; Multidrug resistance-associated protein 9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
94160 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96J65 | |
ABCC12 | |
Recombinant protein corresponding to Human MRP9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only