Learn More
MRPL11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15543-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16824764
  • 4721.67 SEK / 150µL
Estimated Shipment: 22-05-2024
to see stock.
Description
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Sequence analysis identified three transcript variants that encode different isoforms. Pseudogenes corresponding to this gene are found on chromosomes 5q and 12q.
Specifications
Specifications
MRPL11
Polyclonal
Unconjugated
MRPL11
CGI 113, L11MT, MRP L11, MRPL11
Rabbit
Antigen Affinity Chromatography
RUO
293666, 65003, 66419
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5XIE3, Q9CQF0, Q9Y3B7
MRPL11
MRPL11 Fusion Protein Ag7899
150 μL
Primary
Human, Mouse, Rat