Learn More
MRPL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552436
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PAPSLFPAAQ MMNNGLLQQP SALMLLPCRP VLTSVALNAN FVSWKSRTKY TITPVKMRKS GGRDHTGRIR VHGIGGGHKQ RYRMIDFLRF RPEETKSGPF EEKVIQVRYD P Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 71%.
MRPL2 Polyclonal antibody specifically detects MRPL2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
MRPL2 | |
Polyclonal | |
Unconjugated | |
MRPL2 | |
39S ribosomal protein L2, mitochondrial; CGI-22; L2mt; Mitochondrial large ribosomal subunit protein uL2m; mitochondrial ribosomal protein L2; MRP-L14; MRPL2; MRP-L2; ribosomal protein, mitochondrial, L14; Rpml14 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51069 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T653 | |
MRPL2 | |
Recombinant protein corresponding to Human MRPL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only