missing translation for 'onlineSavingsMsg'
Learn More

MRPL36 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16625-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826304

  • 3313.85 SEK / 150µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPL36 encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 2p.
Specifications

Specifications

MRPL36
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9P0J6
BRCA1 interacting protein 1, BRIP1, L36mt, MRP L36, MRPL36, PRPL36, RPMJ
Rabbit
IgG
150 μL
-20°C
Primary
64979
Immunohistochemistry (Paraffin)
Unconjugated
MRPL36
Liquid
MRPL36
MRPL36 Fusion Protein Ag9925
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers