missing translation for 'onlineSavingsMsg'
Learn More

MRPS18B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16139-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16825664

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody recognizes specifically the 29 kDa MRPS18B protein.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S18P family. The encoded protein is one of three that has significant sequence similarity to bacterial S18 proteins. The primary sequences of the three human mitochondrial S18 proteins are no more closely related to each other than they are to the prokaryotic S18 proteins. Pseudogenes corresponding to this gene are found on chromosomes 1q and 2q.
Specifications

Specifications

MRPS18B
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99N84, Q9Y676
C6orf14, HSPC183, HumanS18a, MRP S18 2, MRP S18 b, MRPS18 2, Mrps18 b, MRPS18B, PTD017, S18amt, S18mt b
Rabbit
IgG
150 μL
-20°C
Primary
28973, 294230, 66973
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
MRPS18B
Liquid
Mrps18b
MRPS18B Fusion Protein Ag9198
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers