missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15907-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S2 family. Alternatively spliced transcript variants have been observed for this gene.Specifications
MRPS2 | |
Polyclonal | |
Unconjugated | |
MRPS2 | |
CGI 91, MRP S2, MRPS2, S2mt | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
118451, 362094, 51116 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q924T2, Q9Y399 | |
Mrps2 | |
MRPS2 Fusion Protein Ag8513 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |