missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS27 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550057
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total MRPS27 protein.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that may be a functional partner of the death associated protein 3 (DAP3).Specifications
MRPS27 | |
Polyclonal | |
Unconjugated | |
Mrps27 | |
2610028H14Rik; 28S ribosomal protein S27, mitochondrial; KIAA0264; mitochondrial 28S ribosomal protein S27; mitochondrial ribosomal protein S27; Mitochondrial small ribosomal subunit protein mS27; mrps27; MRP-S27; S27mt | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
218506, 23107 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8BK72, Q92552 | |
Mrps27 | |
Synthesized peptide derived from internal of human MRPS27. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |