missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS30 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18441-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Mitochondrial ribosomes consist of a large 39S subunit and a small 28S subunit, both of which are comprised of multiple mitochondrial ribosomal proteins (MRPs) that are encoded by nuclear genes and are essential for protein synthesis within mitochondria. MRP-S30 (mitochondrial ribosomal protein S30), also known as PDCD9 (programmed cell death protein 9), is a 439 amino acid protein that localizes to the mitochondrion, where it exists as a component of the 28S ribosomal subunit and works in conjunction with other MRPs to mediate protein synthesis. MRP-S30 is expressed in kidney, liver, heart and skeletal muscle. The gene encoding MRP-S30 maps to human chromosome 5, which contains 181 million base pairs and comprises nearly 6% of the human genome. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.Specifications
MRPS30 | |
Polyclonal | |
Unconjugated | |
MRPS30 | |
MRP S30, MRPS30, PAP, PDCD9, S30mt | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10884, 294767, 59054 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9D0G0, Q9NP92 | |
Mrps30 | |
MRPS30 Fusion Protein Ag13271 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |