missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS31 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16288-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. The 28S subunit of the mammalian mitoribosome may play a crucial and characteristic role in translation initiation. This gene encodes a 28S subunit protein that has also been associated with type 1 diabetes; however, its relationship to the etiology of this disease remains to be clarified. Pseudogenes corresponding to this gene have been found on chromosomes 3 and 13.Specifications
MRPS31 | |
Polyclonal | |
Unconjugated | |
MRPS31 | |
Imogen 38, IMOGN38, MRP S31, MRPS31, S31mt | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10240, 290850, 57312 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B0BN56, Q61733, Q92665 | |
Mrps31 | |
MRPS31 Fusion Protein Ag9434 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |