Läs mer
MRS 1754, MedChemExpress
Produktkod. 16468587
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16468587 5 mg
16423406 25 mg
16405968 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16468587 Leverantör Medchem Express Leverantörsnummer HY14121
MedChemExpress MRS 1754 is a selective antagonist radioligand for A2B adenosine receptor with very low affinity for A1 and A3 receptors of both humans and rats.
MRS 1754 is a selective antagonist radioligand for A2B adenosine receptor with very low affinity for A1 and A3 receptors of both humans and rats.
Specifikationer
Kemiskt namn eller material MRS 1754
CAS 264622-58-4
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C26H26N6O4
Kvantitet 5 mg
Löslighetsinformation DMSO : 15 mg/mL (30.83 mM; Need ultrasonic)
LEDER O=C(NC1=CC=C(C#N)C=C1)COC2=CC=C(C(N3)=NC(N(CCC)C(N4CCC)=O)=C3C4=O)C=C2
Molekylvikt (g/mol) 486.52
Formel vikt 486.52
Procent renhet 98.01%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation