Learn More
Abnova™ MRS2L Recombinant Protein
Brand:  Abnova™ H00057380-P01.25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16132643
  • 6845.00 SEK / 25µg
Estimated Shipment: 22-05-2024
to see stock.
Description
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
AAH01028
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
38.61
8
Glutathione Sepharose 4 Fast Flow
25 ug
MECLRSLPCLLPRAMRLPRRTLCALALDVTSVGPPVAACGRRANLIGRSRAAQLCGPDRLRVAGEVHRFRTSDVSQATLASVAPVFTVTKFDKQGNVTSFVFESCDNSRVSSDIRLS
HPT/MGC78523/MRS2L
MRS2
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
57380
MRS2L (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MRS2
Human
Recombinant
Solution