Läs mer
Tocris Bioscience™ MRT 68601 hydrochloride
Potent TBK1 inhibitor
Brand: Tocris Bioscience™ 5067/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MRT 68601 hydrochloride is a potent TBK1 (TANK-binding kinase-1) inhibitor (IC50 = 6 nM). Inhibits the formation of autophagosomes in lung cancer cells.Specifikationer
| N-[3-[[5-Cyclopropyl-2-[[4-(4-morpholinyl)phenyl]amino]-4-pyrimidinyl]amino]propyl]cyclobutanecarboxamide hydrochloride | |
| 1962928-25-1 | |
| 10 mg |