Läs mer
Tocris Bioscience™ MS 21570
Beskrivning
MS 21570 is a GPR171 antagonist (IC50 = 220 nM). Inhibits BigLEN (Cat.No. 6304) mediated increases in [35S]GTPγS binding in hypothalamic membranes and hyperpolarization of basolateral amygdala pyramidal neurons. Reduces anxiety-like behavior and fear conditioning in mice.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Methyl-5-[(phenylmethyl)thio]-1,3,4-thiazadiazol-2-amine |
| CAS | 65373-29-7 |
| Analysprocentintervall | >98% |
| Kvantitet | 50 mg |
| Molekylformel | C10H11N3S2 |
| Formel vikt | Observed MW: 237.34 |