missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A12 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13293-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a cell surface protein found primarily in the apical membrane of colonocytes. Silencing of this gene in colon cancer cells inhibits the proliferation, cell motility, and chemotactic invasion of cells. This gene is part of a cluster of similar genes found on chromosome 11. Two transcript variants encoding different isoforms have been found for this gene.Specifications
MS4A12 | |
Polyclonal | |
Unconjugated | |
MS4A12 | |
Ms4a10, MS4A12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
381213, 54860 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NXJ0 | |
Ms4a12 | |
MS4A12 Fusion Protein Ag3968 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |