missing translation for 'onlineSavingsMsg'
Läs mer

MSI-1436 lactate, MedChemExpress

Produktkod. 16461229
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
5 mg
10 mg
50 mg
Förpackningsstorlek:
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16461229 1 mg -
16475028 10 mg -
16451267 50 mg -
16437757 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16461229 Leverantör Medchem Express Leverantörsnummer HY12219A1mg

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress MSI-1436 lactate is a selective, non-competitive inhibitor of the enzyme protein-tyrosine phosphatase 1B (PTP1B), with an IC50 of 1 μM, 200-fold preference over TCPTP (IC50 of 224 μM).

MSI-1436 lactate is a selective, non-competitive inhibitor of the enzyme protein-tyrosine phosphatase 1B (PTP1B), with an IC50 of 1 μM, 200-fold preference over TCPTP (IC50 of 224 μM).

Specifica

Kemiskt namn eller material MSI-1436 lactate
CAS 1309370-86-2
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C37H72N4O5S.7/2C3H6O3
Kvantitet 1 mg
Synonym Trodusquemine lactate Aminosterol-1436 lactate
Löslighetsinformation DMSO : 54 mg/mL (53.99 mM; Need ultrasonic and warming) ∣0.1 M HCL : 50 mg/mL (49.99 mM; ultrasonic and adjust pH to 4 with HCl)
LEDER NCCCNCCCCNCCCN[C@@H]1C[C@@](C[C@@H](O)[C@]2([H])[C@]3([H])CC[C@@]4(C)[C@@]2([H])CC[C@@]4([C@H](C)CC[C@@H](OS(=O)(O)=O)C(C)C)[H])([H])[C@]3(C)CC1.O=C(O)C(C)O.[3.5]
Molekylvikt (g/mol) 1000.17
Formel vikt 1000.17
Procent renhet 95.0%
Kvalitet Research
Fysisk form Solid
Vedi altri risultati Mostra meno risultati

Research purposes only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.