missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | MSL1 |
---|---|
Concentration | 0.32 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16822845
|
Proteintech
24373-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16812845
|
Proteintech
24373-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Component of histone acetyltransferase complex responsible for the majority of histone H4 acetylation at ′Lys-16′ (H4K16ac) which is implicated in the formation of higher-order chromatin structure. Greatly enhances MSL2 E3 ubiquitin ligase activity, promoting monoubiquitination of histone H2B at ′Lys-34′ (H2BK34Ub). This modification in turn stimulates histone H3 methylation at ′Lys-4′ (H3K4me) and ′Lys-79′ (H3K79me) and leads to gene activation, including that of HOXA9 and MEIS1. In the MSL complex, acts as a scaffold to tether MSL3 and KAT8 together for enzymatic activity regulation.Specifications
MSL1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q68DK7 | |
339287 | |
MSL1 Fusion Protein Ag18024 | |
Primary | |
-20°C | |
MSL1 |
0.32 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
hMSL1, Male specific lethal 1 homolog, Male specific lethal 1 like 1, MSL 1, MSL1, MSL1 like 1, MSL1L1 | |
MSL1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |