missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MSMB (Human) Recombinant Protein
Human MSMB full-length ORF ( AAH05257.1, 1 a.a. - 114 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH05257.1 |
---|---|
Gene ID (Entrez) | 4477 |
Name | microseminoprotein, beta- |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16137961
|
Abnova™
H00004477-P01.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16147961
|
Abnova™
H00004477-P01.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MNVLLGSVVIFATFVTLCNASCYFIPNEGVPGDSTRKCMDLKGNKHPINSEWQTDNCETCTCYETEISCCTLVSTPVGYDKDNCQRIFKKEDCKYIVVEKKDPKKTCSVSEWII
Specifications
AAH05257.1 | |
microseminoprotein, beta- | |
125% SDS-PAGE Stained with Coomassie Blue | |
HPC13, IGBF, MSP, MSPB, PN44, PRPS, PSP, PSP-94, PSP57, PSP94 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
4477 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MSMB | |
GST |