missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MSMB (Human) Recombinant Protein
Human MSMB full-length ORF ( AAH05257.1, 1 a.a. - 114 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004477-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MNVLLGSVVIFATFVTLCNASCYFIPNEGVPGDSTRKCMDLKGNKHPINSEWQTDNCETCTCYETEISCCTLVSTPVGYDKDNCQRIFKKEDCKYIVVEKKDPKKTCSVSEWII
Specifications
AAH05257.1 | |
microseminoprotein, beta- | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MSMB | |
GST |
4477 | |
Wheat germ expression system | |
10 μg | |
HPC13, IGBF, MSP, MSPB, PN44, PRPS, PSP, PSP-94, PSP57, PSP94 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |