missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSRB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14251-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene catalyzes the reduction of methionine sulfoxide to methionine. This enzyme acts as a monomer and requires zinc as a cofactor. Several transcript variants encoding two different isoforms have been found for this gene. One of the isoforms localizes to mitochondria while the other localizes to endoplasmic reticula.Specifications
MSRB3 | |
Polyclonal | |
Unconjugated | |
MSRB3 | |
MSRB3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
253827, 320183, 680036 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BU85, Q8IXL7 | |
Msrb3 | |
MSRB3 Fusion Protein Ag5524 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |