Learn More
Descrizione
Thermo Scientific™ MSSA provides a continuous window into the hydrometallurgical process which allows operators or an automatic control system to increase plant efficiency, quality and recovery. The Thermo Scientific Multi-Stream Solution Analyzer, MSSA, provides continuous online analysis of solution streams and is capable ofmeasuring elements as low as sulphur in the periodic table. This provides costeffective, real-time analysis of key elements and other variables from sampledstream lines in hydrometallurgical operations such as SX/EW and refineries.
Key Features
- Simultaneously measures measure 8 elements
- Receives between 1 and 16 streams per analyzer
- Energy Dispersive XRF technique of analysis
- Stainless-steel frame and construction
- Stainless-steel or polypropylene wetted parts
- Simple, low maintenance design
- De-multiplexing of streams available if required
- Metallurgical shift sampling available
- Flush capabilities for scaling solutions
Specific Benefits
- Monitors the transfer of metal species through different stages of the plant, viz: pregnant liquor solution, organic and electrolytes.
- Monitors contaminant species in copper SX/EW in aqueous streams
- Automated rectifiers in the EW plant
- Optimized electrical energy use in the EW plant by minimizing peak power loads
- Optimized current density based on metal concentration in the electrolyte to improve cathode quality
- Automated reagent dosing
- Manpower savings due to the decrease in control samples analyzed by the laboratory
Ordering Alerts:
A single-stream analyzer (SSSA) is also available; for information contact your Thermo Scientific Sales Representative.
Specifica
Specifica
| Type | MSSA Multi-Stream Solution Analyzer |
| Description | Multi-Stream Solution Analyzer, SamStat-30 Slurry Sampling Station (For details contact your Sales Representative) |
| Certifications/Compliance | Manufactured in an ISO 9001:2000 certified facility. |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.