missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MST1R (RON) Recombinant Human Protein
MST1R (RON) Recombinant Human Protein
Brand: Invitrogen PV4314
Additional Details : Weight : 0.05250kg
Description
MST1R (RON) is a receptor tyrosine kinase involved in cell proliferation and motility.Specifications
NP_002438 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
74.8kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
MST1R (RON) | |
MST1R (RON) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL AXLtide peptide substrate (KKSRGDYMTMQIG) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 983-1400 | |
1730nmoles of phosphate transferred to AXLtide peptide substrate (KKSRGDYMTMQIG) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1.67μg/mL | |
GST Tag | |
Kinase and inhibitors | |
90% by SDS-PAGE |
0.41mg/mL | |
114294, 4486 | |
MST1R (RON) Recombinant Human Protein | |
–80°C | |
1 Each | |
MST1R (RON) Recombinant Human Protein | |
MET (HGFR) | |
MST1R (RON) Recombinant Human Protein | |
Human | |
Baculovirus | |
Receptor tyrosine kinases |
For Research Use Only. Not for use in diagnostic procedures.