missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
MSX1/MSX2 Polyclonal Antibody, Invitrogen™
Beskrivning
Predicted to react with Mouse and Rat samples.
Drosophila,muscle segment (msh) homolog 1,homeo domain encoding gene,inhibiting MYOD1 expression,highly expressed in dental mesenchyme during critical bud stage,involved in epithelial-mesenchymal signaling in many organs,and in the pathogenesis of cleft lip and palate,interacting with MSX2 in mouse limb bud patterning .This gene encodes a member of the muscle segment homeobox gene family. The encoded protein functions as a transcriptional repressor during embryogenesis through interactions with components of the core transcription complex and other homeoproteins. It may also have roles in limb-pattern formation, craniofacial development, particularly odontogenesis, and tumor growth inhibition.Tissue specificity: Expressed in the developing nail bed mesenchyme.
Specifikationer
Specifikationer
| Antigen | MSX1/MSX2 |
| Användningsområden | Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gen | MSX1 |
| Genaccessionsnr. | P52953, P13297, Q03358, P28360, P35548 |
| Gensymboler | MSX1, MSX2 |
| Värd art | Rabbit |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?