missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ MT-ND4L Polyclonal Antibody

Beskrivning
MT-ND4L Polyclonal Antibody for Western Blot
NADH:ubiquinone oxidoreductase (complex I) is an extremely complicated multiprotein complex located in the inner mitochondrial membrane. Human complex I is important for energy metabolism because its main function is to transport electrons from NADH to ubiquinone, which is accompanied by translocation of protons from the mitochondrial matrix to the intermembrane space. Human complex I appears to consist of 41 subunits. A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7), while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 4L (ND4L) is most likely a component of the hydrophobic protein fragment of Complex I. Mutations in the gene encodiing for ND4 are implicated in Leber hereditary optic neuropathy, a rare condition that can cause loss of central vision.
Specifikationer
Specifikationer
| Antigen | MT-ND4L |
| Användningsområden | Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gen | MT-ND4L |
| Genaccessionsnr. | P03901 |
| Gene Alias | mitochondrially encoded NADH 4L dehydrogenase; MTND4L; mt-Nd4l; NADH dehydrogenase subunit 4L; NADH4L; NADH-ubiquinone oxidoreductase chain 4L; Nd4l |
| Gensymboler | MT-ND4L |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?