missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MT1A (Human) Recombinant Protein
Human MT1A full-length ORF ( AAH29475.1, 1 a.a. - 61 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004489-P01.25ug
This item is not returnable.
View return policy
Description
- Sequence: MDPNCSCATGGSCTCTGSCKCKECKCNSCKKSCCSCCPMSCAKCAQGCICKGASEKCSCCA
Specifications
AAH29475.1 | |
metallothionein 1A | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MT1A | |
GST |
4489 | |
Wheat germ expression system | |
25 μg | |
MGC32848, MT1, MT1S, MTC | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |