missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MT2A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20809-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Metallothionein (MT) is a sulfhydryl- and cysteine-rich protein found in microorganisms, plants and all invertebrate and vertebrate animals. Metallothioneins are a group of ubiquitous low-molecular-weight proteins that have functional roles in cell growth, repair and differentiation. They are implicated primarily in metal ion detoxification, in that they are essential for the protection of cells against the toxicity of cadmium, mercury and copper. Metallothionein, as an acute phase or stress-response protein and free radical scavenger, is related to inflammation and cellular protection from reactive forms of oxygen, ionizing radiation, pharmacological agents and mutagens. Metallothioneins are known to be broadly expressed in heart, liver, kidney, breast and testis tissue.Specifications
MT2A | |
Polyclonal | |
Unconjugated | |
MT2A | |
CES1, Metallothionein 2, metallothionein 2A, Metallothionein II, MT 2, MT II, MT2, MT2A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4502 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P02795 | |
MT2A | |
MT2A Fusion Protein Ag14761 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |