missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTFR1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555512
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: CSASPECCKP EHKAACSSSE EDDCVSLSKA SSFADMMGIL KDFHRMKQSQ DLNRSLLKEE DPAVLISEVL RRKFALKEED Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.
MTFR1L Polyclonal antibody specifically detects MTFR1L in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
MTFR1L | |
Polyclonal | |
Unconjugated | |
MTFR1L | |
FAM54B; family with sequence similarity 54 member B; family with sequence similarity 54, member B; HYST1888; mitochondrial fission regulator 1 like; mitochondrial fission regulator 1-like; MST116; MSTP116; MTFR1L; protein FAM54B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56181 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H019 | |
MTFR1L | |
Recombinant protein corresponding to Human MTFR1L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only