Learn More
MTH1 Mouse anti-Human, Unconjugated, Clone: 2D7G4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67443-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The MTH1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MTH1. This antibody recognizes human antigen. The MTH1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
MTH1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
2D7G4 | |
Unconjugated | |
20 μL | |
4521 | |
IgG1 |