missing translation for 'onlineSavingsMsg'
Learn More

MTMR6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13684-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16822034

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Phosphatase that acts on lipids with a phosphoinositol headgroup. Acts as a negative regulator of KCNN4/KCa3.1 channel activity in CD4+ T-cells possibly by decreasing intracellular levels of phosphatidylinositol 3 phosphatase. Negatively regulates proliferation of reactivated CD4+ T-cells.
Specifications

Specifications

MTMR6
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8VE11, Q9Y217
MTMR6, myotubularin related protein 6
Rabbit
IgG
20 μL
-20°C
Primary
219135, 305935, 9107
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MTMR6
Liquid
Mtmr6
MTMR6 Fusion Protein Ag4551
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Rat, Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers