missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mTOR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20657-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The antibody is specific to MTOR.
Mammalian target of rapamycin (mTOR) is a serine/threonine kinase that plays a key role in cell growth, cell proliferation, and protein synthesis. mTOR mediates phosphoinositide 3-kinase and Akt/PKB signaling, resulting in phosphorylation of 4EBP1, and initiation of mRNA translation. A second pathway involves regulation of ribosomal S6 kinase, which affects ribosome biogenesis and translation elongation. Mutations affecting the gene results in Smith-Kingsmore syndrome.Specifications
mTOR | |
Polyclonal | |
Unconjugated | |
MTOR | |
FRAP, FRAP1, FRAP2, Mammalian target of rapamycin, MTOR, RAFT1, Rapamycin target protein 1, RAPT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2475, 56717, 56718 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P42345, P42346, Q9JLN9 | |
Mtor | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |