missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mucin 3 (MUC3) Ab-2 Mouse Monoclonal Antibody, Epredia™
Epredia™ Mucin 3 (MUC3) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
1499.40 SEK - 6951.30 SEK
Specifications
Antigen | Mucin 3 (MUC3) Ab-2 |
---|---|
Clone | 1143/B7 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
12610097
|
Epredia
MS-1734-P1 |
200μg/mL | 500 μL |
5440.00 SEK
500µL |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12620097
|
Epredia
MS-1734-P1ABX |
1mg/mL | 100 μL |
6260.00 SEK
100µL |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12690087
|
Epredia
MS-1734-P |
200μg/mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12600097
|
Epredia
MS-1734-P0 |
200μg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12630097
|
Epredia
MS-1734-PABX |
1mg/mL | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Mucin 3 (MUC3) is a glycoprotein. Distributed in colon and rectum, and to a lesser extent in breast, lung and salivary gland tissues.Host Species: Mouse
Clone: 1143/B7
Isotype: IgG2a
Species Reactivity: Human. Others not known.
Epitope: Tandem repeats of the human MUC3
Immunogen: MUC3 tandem repeat peptide
Molecular Weight: High
Positive Control: Colon CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Mucin 3 (MUC3) Ab-2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer and Tumor Biology | |
MUC3 tandem repeat peptide | |
Primary |
1143/B7 | |
Monoclonal | |
Concentrated | |
RUO | |
Human | |
IgG2a |