missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPML3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564006
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KGYMDRMDDT YAVYTQSDVY DQLIFAVNQY LQLYNVSVGN HAY Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.
TRPML3 Polyclonal antibody specifically detects TRPML3 in Human samples. It is validated for Immunohistochemistry.Specifications
TRPML3 | |
Polyclonal | |
Unconjugated | |
MCOLN3 | |
6720490O21Rik; FLJ11006; FLJ36629; MCLN3; Mcoln3; Mcoln3 mucolipin 3; MGC124245; MGC124246; MGC71509; mucolipin 3; mucolipin-3; Transient receptor potential channel mucolipin 3; TRPML3; TRP-ML3; Va; varitint-waddler | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55283 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TDD5 | |
MCOLN3 | |
Recombinant protein corresponding to Human Mucolipin 3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only