Learn More
MUDENG Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26567-1-AP-20UL
 View more versions of this product
Product Code. 16884985
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
As part of AP-5, a probable fifth adaptor protein complex it may be involved in endosomal transport.
Specifications
Specifications
MUDENG
Polyclonal
Unconjugated
AP5M1
AP-5 complex subunit mu-1, AP5M1, C14orf108, MuD, MUDENG
Rabbit
Antigen Affinity Chromatography
RUO
55745
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9H0R1
AP5M1
MUDENG Fusion Protein Ag23486
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MUDENG Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated