Learn More
Nabertherm™ Muffle Furnace with Flap Door
The muffle furnaces L 1/12 - LT 40/12 are the right choice for daily laboratory use. These models stand out for their excellent workmanship, advanced and attractive design, and high level of reliability. The muffle furnaces come equipped with lift door at no extra charge.
40010.00 SEK - 69190.00 SEK
Specifications
Description Flap door
View More Specs
Products 11
Product Code Brand Capacity (Metric) Controller Type Temperature (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Controller Type Temperature (Metric) Price Quantity & Availability  
15466682
View Documents
Nabertherm™
L-052K2TN2
5 L C450 1200°C 42800.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
06-11-2020
Add to basket
15486682
View Documents
Nabertherm™
L-052S3RN3
5 L B410 1300°C 40010.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
27-11-2020
Add to basket
15496682
View Documents
Nabertherm™
L-052S3TN3
5 L C450 1300°C 46340.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
27-11-2020
Add to basket
15436702
View Documents
Nabertherm™
L-092K2TN2
9 L C450 1200°C 48485.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
06-11-2020
Add to basket
15456712
View Documents
Nabertherm™
L-152S3RN3
15 L B410 1300°C 48700.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
27-11-2020
Add to basket
15466712
View Documents
Nabertherm™
L-152S3TN3
15 L C450 1300°C 55030.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
27-11-2020
Add to basket
15436722
View Documents
Nabertherm™
L-242K1RN1
24 L B410 1100°C 52135.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
27-11-2020
Add to basket
15456722
View Documents
Nabertherm™
L-242K2RN2
24 L B410 1200°C 59965.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
27-11-2020
Add to basket
15466722
View Documents
Nabertherm™
L-242K2TN2
24 L C450 1200°C 66290.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
27-11-2020
Add to basket
15436732
View Documents
Nabertherm™
L-402K1TN1
40 L C450 1100°C 60930.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
27-11-2020
Add to basket
15456732
View Documents
Nabertherm™
L-402K2TN2
40 L C450 1200°C 69190.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
27-11-2020
Add to basket
Description
Description
  • Door: Flap door
  • tabletop model
Specifications
Specifications
Flap door