missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16133-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
E3 ubiquitin-protein ligase that plays a role in the control of mitochondrial morphology. Promotes mitochondrial fragmentation and influences mitochondrial localization. Inhibits cell growth. When overexpressed, activates JNK through MAP3K7/TAK1 and induces caspase-dependent apoptosis. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates.Specifications
MUL1 | |
Polyclonal | |
Unconjugated | |
MUL1 | |
C1orf166, GIDE, MAPL, MUL1, MULAN, RING finger protein 218, RNF218, RP11 401M16.2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
298576, 68350, 79594 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8VCM5, Q969V5 | |
MUL1 | |
MUL1 Fusion Protein Ag9119 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |