missing translation for 'onlineSavingsMsg'
Learn More

Multi tags Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24147-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16822695

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody recognize multi tags include: GST tag, 6xHIS tag, DDDDK tag, MYC tag and V5 tag.

Specifications

Specifications

Multi tags
0.49 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
6x HIS tag, DDDDK tag, GST tag, MYC tag
Multi tags Fusion Protein Ag21457
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Western Blot
Unconjugated
Liquid
Rabbit
IgG
20 μL
-20°C
Primary
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers