missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Multi tags Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | Multi tags |
---|---|
Concentration | 0.49 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16822695
|
Proteintech
24147-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16812695
|
Proteintech
24147-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody recognize multi tags include: GST tag, 6xHIS tag, DDDDK tag, MYC tag and V5 tag.
Specifications
Multi tags | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
6x HIS tag, DDDDK tag, GST tag, MYC tag | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
0.49 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Multi tags Fusion Protein Ag21457 | |
Primary | |
-20°C |