Learn More
Thermo Scientific™ Multidrop™ Combi SMART Reagent Dispenser Accessories
Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi SMART Reagent Dispenser Accessories.
Brand:  Thermo Scientific™ N15138
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 15451825
  • 28100.26 SEK / Pack of 5
Estimated Shipment
11-01-2019
Add to basket
Description
Description
  • Cat. No. N15133 to N15139 are to be used with the Multidrop Combi with SMART2 option instrument with serial numbers 836-2846SM onwards
  • Cat. No. 24073292, 24073001, 24073297, 24073002, 24072675, 24072678 and 24072676 are to be used with the Multidrop Combi with SMART option instrument with serial numbers up to 836-2845SM
Specifications
Specifications
SMART 2 standard tube dispensing cassette 5-pack, new RFID tag
50 times at 121°C, 1 bar, 20 min.
5 to 2500µL
<7mL
Multidrop Combi with SMART2 option instrument with serial numbers 836-2846SM onwards
Tubing: 1.3mm; Tip: 0.5mm
Tip: PP; Tubing: Silicone
Videos