Läs mer
Mupirocin Calcium Salt, TRC
Produktkod. 30467766
Change view
Kvantitet:
100 mg
250 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30467766 250 mg
30481848 2.5 g
30473991 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30467766 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM794005250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 115074-43-6
Molekylformel 2 C26 H43 O9 . Ca . 2 H2 O
Molekylvikt (g/mol) 1075.34
Synonym Mupirocin calcium, Calcium bis[9-[[(2E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5[[(2S,3S)-3-[(1S,2S)-2-hydroxy-1methylpropyl]oxiranyl]methyl]tetrahydro-2H-pyran-2-yl]-3-methylbut-2-enoyl]oxy]nonanoate] dihydrate
IUPAC-namn calcium;9-[(E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoyl]oxynonanoate;dihydrate
LEDER O.O.[Ca+2].C[C@H](O)[C@H](C)[C@@H]1O[C@H]1C[C@H]2CO[C@@H](C\C(=C\C(=O)OCCCCCCCCC(=O)[O-])\C)[C@H](O)[C@@H]2O.C[C@H](O)[C@H](C)[C@@H]3O[C@H]3C[C@H]4CO[C@@H](C\C(=C\C(=O)OCCCCCCCCC(=O)[O-])\C)[C@H](O)[C@@H]4O
Specifikationer
Kemiskt namn eller material Mupirocin Calcium
Kvantitet 250 mg
InChI formel InChI=1S/2C26H44O9.Ca.2H2O/c2*1-16(13-23(30)33-11-9-7-5-4-6-8-10-22(28)29)12-20-25(32)24(31)19(15-34-20)14-21-26(35-21)17(2)18(3)27;;;/h2*13,17-21,24-27,31-32H,4-12,14-15H2,1-3H3,(H,28,29);;2*1H2/q;;+2;;/p-2/b2*16-13+;;;/t2*17-,18-,19-,20-,21-,24+,25-,26-;;;/m00.../s1
Formel vikt 1074.5651