Läs mer
Muraglitazar, TRC
Produktkod. 30480570
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480570 5 mg
30469849 1 mg
30472370 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30480570 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM7965005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 331741-94-7
Molekylformel C29 H28 N2 O7
Molekylvikt (g/mol) 516.54
IUPAC-namn 2-[(4-methoxyphenoxy)carbonyl-[[4-[2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethoxy]phenyl]methyl]amino]acetic acid
LEDER COc1ccc(OC(=O)N(CC(=O)O)Cc2ccc(OCCc3nc(oc3C)c4ccccc4)cc2)cc1
Specifikationer
Kemiskt namn eller material Muraglitazar
Kvantitet 5 mg
InChI formel InChI=1S/C29H28N2O7/c1-20-26(30-28(37-20)22-6-4-3-5-7-22)16-17-36-24-10-8-21(9-11-24)18-31(19-27(32)33)29(34)38-25-14-12-23(35-2)13-15-25/h3-15H,16-19H2,1-2H3,(H,32,33)
Formel vikt 516.1897