Learn More
Gibco™ Murine Dendritic Cell Cytokine Package, PeproTech®
Product Code. 17823423
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Product Code. Quantity Unit Size
17823423 1 Kit Each
1 options
Product Code. 17823423 Supplier Gibco™ Supplier No. MDC
  • ONLINE EXCLUSIVE PRICE
    9340.00 SEK
    8499.40 SEK / Paket med 1
    Save 840.60 SEK
    9% Av
 Restricted Item
Beräknad leverans: 04-12-2026
The Murine Dendritic Cell Cytokine Package contains the key components required for the culturing of mouse dendritic cells.
IL-4 is a pleiotropic cytokine that regulates diverse T- and B-cell responses including cell proliferation, survival, and gene expression. Produced by mast cells, T cells, and bone marrow stromal cells, IL-4 regulates the differentiation of naive CD4+ T cells into helper Th2 cells.
GM-CSF is a hematopoietic growth factor that stimulates the development of neutrophils and macrophages and promotes the proliferation and development of early erythroid megakaryocytic and eosinophilic progenitor cells.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Functional Assay
Product Type Dendritic Cell Cytokine Package
Quantity 1 Kit
Content And Storage 100 μg Murine IL-4 (Cat. No. 214-14-100 UG), 100 μg Murine IL-4 (Cat. No. 214-14-100 UG) 100 μg Murine GM-CSF (Cat. No. 315-03-100 UG), 100 μg Murine GM-CSF (Cat. No. 315-03-100 UG)
Species Mouse