Läs mer
Honeywell™ Mutex E Light Protective Clothing
Produktkod. 15683346
Klicka för att se tillgängliga alternativ
Storlek:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Förpackningsstorlek:
Varje
Produktkod. 15683346
Brand: Honeywell™ 4500101S
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Breathable, single-use protective clothing designed to protect against particle contamination and chemical splashes.
  • Antistatic
  • Silicone and latex free
  • Excellent ergonomics and fitting
    • 3-piece hood
    • Special crotch design
    • Outside flap and dual slider sipper
    • Breathability
    • 3-layer polypropylene
TRUSTED_SUSTAINABILITY
Specifikationer
Produkttyp Chemical-resistant Coveralls
Storlek Small
Kön Unisex
Material 3-layer Polypropylene
Färg White
Skyddsklass Protection type: 5, 6
Tjocklek 56 gsm