Learn More
Honeywell™ Mutex E Light Protective Clothing
Breathable, single-use protective clothing designed to protect against particle contamination and chemical splashes.
61.76 SEK - 115.00 SEK
Specifications
Product Type Chemical-resistant Coveralls
Gender Unisex
Color White
Protection Class Protection type: 5, 6
Thickness 56 gsm
View More Specs
Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15683346
View Documents
Honeywell™
4500101-S
Small This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15673346
View Documents
Honeywell™
4500101-M
Medium This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15663346
View Documents
Honeywell™
4500101-L
Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15693346
View Documents
Honeywell™
4500101-XL
X-Large
108.00 SEK
Each
Only null left
Estimated Shipment: 25-09-2025
Add to basket
15603356
View Documents
Honeywell™
4500101-XXL
2X-Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15613356
View Documents
Honeywell™
4500101-XXXL
3X-Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Antistatic
  • Silicone and latex free
  • Excellent ergonomics and fitting
    • 3-piece hood
    • Special crotch design
    • Outside flap and dual slider sipper
    • Breathability
    • 3-layer polypropylene
Specifications
Specifications
Chemical-resistant Coveralls
White
56 gsm
Unisex
Protection type: 5, 6
Documents
Product Certifications
Product Content Correction
Product Title
Honeywell™ Mutex E Light Protective Clothing