missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myc Epitope Tag Antibody (9E10), Alexa Fluor™ 488, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP2-81018AF488
This item is not returnable.
View return policy
Description
Myc Epitope Tag Monoclonal antibody specifically detects Myc Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).Specifications
Myc Epitope Tag | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Human | |
Protein A purified | |
RUO | |
Epitope sequence, EQKLISEEDL, located in aa 410-419 of human c-myc protein. | |
Purified | |
Epitope Tag |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
90000000000 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
BHLHE39, Class E basic helix-loop-helix protein 39 (bHLHe39), Myc proto-oncogene protein, Proto-oncogene c-Myc, Transcription factor p64 | |
A synthetic peptide corresponding to aa 408-439 from C-terminus of human myc. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |