Läs mer
Myclobutanil, TRC
Produktkod. 17933185
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17933185 250 mg
30132887 1 g
17902235 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17933185 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM831400250MG
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 88671-89-0
Kemiskt namn eller material Myclobutanil
Formel vikt 288.1142
InChI formel InChI=1S/C15H17ClN4/c1-2-3-8-15(9-17,10-20-12-18-11-19-20)13-4-6-14(16)7-5-13/h4-7,11-12H,2-3,8,10H2,1H3
IUPAC-namn 2-(4-chlorophenyl)-2-(1,2,4-triazol-1-ylmethyl)hexanenitrile
Molekylformel C15 H17 Cl N4
Molekylvikt (g/mol) 288.78
Procent renhet >95%
Anteckningar om renhetsgrad HPLC
LEDER CCCCC(Cn1cncn1)(C#N)c2ccc(Cl)cc2
Synonym 1H-1,2,4-Triazole-1-propanenitrile, α-butyl-α-(4-chlorophenyl)-, α-Butyl-α-(4-chlorophenyl)-1H-1,2,4-triazole-1-propanenitrile, (±)-Myclobutanil, Deccotanil, Eagle, Fungiben, HOE 39304F, Laredo 25EC, Laredo 25EW, Myclobutanil, Mycloss Xtra, Nova, Nova (pesticide), Nova 40W, Nova W, Nu-Flow M, RH 3866, Rally, Spectracide Immunox Multi-Purpose Fungicide, Systhane, Systhane 12E, Systhane 40W, Systhane 6 Flo